Learn More
Aqua Solutions Sodium Standard 100 ppm in 3.5% Nitric Acid NIST Traceable (500ML)
SDP
Catalog No. NC9648006
Encompass_Preferred
Sodium Standard 100 ppm in 3.5% Nitric Acid NIST Traceable (500ML)
Catalog No. NC9648006 Supplier Aqua Solutions Supplier No. 8751500ML
Sodium Standard 100 ppm in 3.5% Nitric Acid NIST Traceable (500ML)