Learn More
Sodium stearate
Description
Used in adhesives and sealants, laundry and dishwashing products, plastic and rubber products. Used as surface active agents. It is the gelling agent for deodrant sticks. Used as waterproofing additives and ointments.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Odor | Weak |
| Quantity | 250 g |
| Merck Index | 14,8678 |
| Solubility Information | Slowly soluble in cold water. |
| Formula Weight | 306.46 |
| Physical Form | Powder |
| Chemical Name or Material | Sodium stearate |
Safety and Handling
| Hazard Category | H315-H319-H500 |
| Precautionary Statement | P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362 |
| EINECSNumber | 212-490-5 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.