Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Stearoyllactate (so called), TCI America™
SDP

Catalog No. S084625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S084625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S084625G Supplier TCI America Supplier No. S084625G

Specifications

Color White-Yellow
Quantity 25 g
Synonym Stearoyllactic Acid Sodium Salt
Physical Form Crystalline Powder
Chemical Name or Material Sodium Stearoyllactate (so called)
EINECSNumber (7)-2115
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.