Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sodium Succinate Dibasic Hexahydrate

Catalog No. S2378100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
250 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S2378100G 1 kg
S2378500G 100 g
S23781KG 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S2378100G Supplier Sigma Aldrich Supplier No. S2378100G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

ReagentPlus is a registered trademark of Sigma-Aldrich Co. LLC

Specifications

CAS 6106-21-4
Molecular Formula C4H4Na2O4 -+ 6H2O
Linear Formula NaOOCCH2CH2COONa -+ 6H2O
MDL Number MFCD00149117
Quantity 1 kg
Synonym Butanedioic acid disodium salt; Succinic acid disodium salt
Molecular Weight (g/mol) 270.14 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.