Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sodium Succinate Dibasic Hexahydrate

Catalog No. S23781KG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
250 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S23781KG 250 g
S2378500G 100 g
S2378100G 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S23781KG Supplier Sigma Aldrich Supplier No. S23781KG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

ReagentPlus is a registered trademark of Sigma-Aldrich Co. LLC

Specifications

CAS 301-10-0
Density 1.251 g/mL at 25°C
Molecular Formula C16H30O4Sn
Linear Formula [CH3(CH2)3CH(C2H5)CO2]2Sn
MDL Number MFCD00002676
Quantity 250 g
Synonym Butanedioic acid disodium salt; Succinic acid disodium salt
Molecular Weight (g/mol) 405.12 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.