Learn More
Apexbio Technology LLC SODIUM-SULFADIAZINE-5G
SDP
Catalog No. 5000568863
Encompass_Preferred
Sodium sulfadiazine (CAS No 547-32-0) is a sulfonamide antibiotic primarily used in biomedical research categories focusing on antimicrobial mechanisms It acts by inhibiting bacterial dihydropteroate synthase disrupting folic acid synthesis essential for bacterial growth Its efficacy is often observed in vitro within the nanomolar to low micromolar range against a range of bacterial strains Typical applications include studies on bacterial resistance cell model assays and animal models to evaluate antimicrobial efficacy and pharmacokinetics It is also employed in drug screening to assess interactions with other antibiotics Experimental concentrations typically depend on the specific research design Sodium sulfadiazine s role in research underscores its importance in understanding drug-pathogen interactions and potential therapeutic strategies
Catalog No. 50-005-68863 Supplier Apexbio Technology LLC Supplier No. C64735000
Sodium sulfadiazine (CAS No 547-32-0) is a sulfonamide antibiotic primarily used in biomedical research categories focusing on antimicrobial mechanisms It acts by inhibiting bacterial dihydropteroate synthase disrupting folic acid synthesis essential for bacterial growth Its efficacy is often observed in vitro within the nanomolar to low micromolar range against a range of bacterial strains Typical applications include studies on bacterial resistance cell model assays and animal models to evaluate antimicrobial efficacy and pharmacokinetics It is also employed in drug screening to assess interactions with other antibiotics Experimental concentrations typically depend on the specific research design Sodium sulfadiazine s role in research underscores its importance in understanding drug-pathogen interactions and potential therapeutic strategies