Learn More
Strem, An Ascensus Company CAS# 7757-82-6. 1kg. Sodium sulfate, anhydrous, 99+% (ACS). MFCD00003504
SDP
Catalog No. 5090113816
Encompass
CAS# 7757-82-6. 1kg. Sodium sulfate, anhydrous, 99+% (ACS). MFCD00003504. Molecular Weight: 142.04. Molecular Formula: Na2SO4. Color/form: white pwdr. Strem# 93-1068. http://www.strem.com/catalog/v/93-1068/
Catalog No. 50-901-13816 Supplier Strem, An Ascensus Company Supplier No. 9310681000G
CAS# 7757-82-6. 1kg. Sodium sulfate, anhydrous, 99+% (ACS). MFCD00003504. Molecular Weight: 142.04. Molecular Formula: Na2SO4. Color/form: white pwdr. Strem# 93-1068. http://www.strem.com/catalog/v/93-1068/