Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Sulfate USP, Macron Fine Chemicals™

Catalog No. MK8028212 Shop All Avantor Products
Change view
Click to view available options
Poly; 2.5kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
MK-8028-212 Poly; 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MK-8028-212 Supplier Avantor Supplier No. 802806
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 7757-82-6
Quantity 2.5 kg
Molecular Formula Na2O4S
MDL Number MFCD00003504
Synonym sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1
InChI Key PMZURENOXWZQFD-UHFFFAOYSA-L
SMILES [Na+].[Na+].[O-]S([O-])(=O)=O
IUPAC Name disodium;sulfate
Molecular Weight (g/mol) 142.04
PubChem CID 24436
ChEBI CHEBI:32149
Formula Weight 142.04
Grade USP
Packaging Poly
Chemical Name or Material Sodium Sulfate, Anhydrous
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.