Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Sulfathiazole, Sesquihydrate, 99%, Spectrum™ Chemical
SDP

Catalog No. 18606839 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 g
1 kg
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-839 100 g Poly Bottle
18-606-841 1 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-606-839 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S1462100GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

S1462, C9H8N3NaO2S2·1.5H2O

Spectrum™ Chemical Sodium Sulfathiazole, Sesquihydrate , an organosulfur compound, can be used as a short-acting sulfa drug and as an oral and topical antimicrobial, Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Specifications

CAS Max % 1
Quantity 100 g
Percent Purity 99%
Grade Reagent
Loss on Drying 0.09
Packaging Poly Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.