Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Sulfide Hydrate (Flakes/Technical), Fisher Chemical

Catalog No. S426212 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
2.5 kg
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S426-212 2.5 kg Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S426-212

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1313-82-2 , 7732-18-5
Molecular Formula Na2S
Molecular Weight (g/mol) 78.04
MDL Number MFCD00003498
InChI Key GRVFOGOEDUUMBP-UHFFFAOYSA-N
Synonym sodium sulfide, sodium sulfide na2s, disodium monosulfide, na2s, disodium sulphide, sodium sulfide van, hsdb 772, sodium sulfide, anhydrous dot, sodum sulfuret, na2-s
PubChem CID 237873
IUPAC Name disodium sulfanediide
SMILES [Na+].[Na+].[S--]

Specifications

Color Yellow
Physical Form Solid
Quantity 2.5 kg
Formula Weight 78.04
Grade Technical
Identification Pass Test
Packaging Glass Bottle
Chemical Name or Material Sodium Sulfide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.