Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sodium Sulfide Hydrate

Catalog No. 1346825KGR Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-468-25KGR 2.5 kg
13-468-1KGR 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger-Danger
Catalog No. 13-468-25KGR Supplier Sigma Aldrich Supplier No. 134682.5KGR
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger-Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

May contain up to 40% water.Nucleophile and reducing agent derived from H2S; reagent for the synthesis of thiols, thioethers, and acyclic sulfides, di-, and polysulfides; reduces aromatic nitro compounds to amines; reduces sulfoxides to sulfides.

Specifications

CAS 27610-45-3
Quantity 2.5 kg
Molecular Formula Na2S -+ xH2O
Linear Formula Na2S -+ xH2O
MDL Number MFCD00003498
Molecular Weight (g/mol) 78.04 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.