Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sodium sulfide nonahydrate

Catalog No. 431648250G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-164-8250G 250 g
43-164-850G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 43-164-8250G Supplier Sigma Aldrich Supplier No. 431648250G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Sodium sulfide nonahydrate (Na2S.9H2O) can be reacted withα,-d-dibromo and/orα,-d-dichloroalkane for the synthesis of polyalkylene sulfide. It can be used as a reducing agent for the removal of nitrogen oxide from the exhaust gas. Na2S.9H2O can also be used as an inexpensive sulfur source for the synthesis of heterocyclic thiochromes.

Specifications

CAS 1313-84-4
Quantity 250 g
Molecular Formula Na2S -+ 9H2O
Linear Formula Na2S-+9H2O
MDL Number MFCD00149184
Synonym Disodium sulfide nonahydrate; Sodium sulfide enneahydrate
Molecular Weight (g/mol) 240.18
Percent Purity ≥99.99% trace metals basis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.