Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Sulfide Nonahydrate (Crystalline/Certified ACS), Fisher Chemical

Catalog No. S425500 Shop All Fisherbrand Products
Change view
Click to view available options
Poly Bottle; 500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S425-500 Poly Bottle; 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S425-500

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This chemical is deliquescent

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 50°C
Color Yellow
CAS 1313-84-4
Physical Form Solid
Quantity 500 g
Assay Percent Range ≥98.0 %
Molecular Formula H18Na2O9S
MDL Number MFCD00149184
Synonym Sodium Monosulfide
InChI Key ZGHLCBJZQLNUAZ-UHFFFAOYSA-N
SMILES O.O.O.O.O.O.O.O.O.[Na+].[Na+].[S--]
IUPAC Name disodium nonahydrate sulfanediide
Molecular Weight (g/mol) 240.17
Formula Weight 240.18
Percent Purity ≥98.0%
Grade Certified ACS
Identification Pass Test
Packaging Poly Bottle
Chemical Name or Material Sodium Sulfide Nonahydrate
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Toxic in contact with skin. Causes burns by all exposure routes. Contact with acids liberates toxic gas. Very toxic to aquatic organisms. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici.

NFPA

Health:3
Flammability:1
Instability:1
DOTInformation DOT Class 8, : Corrosive
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.