Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Sulfite, 1.00 M, 2.00 N, Ricca Chemical
SDP

Catalog No. R783200020F Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
20 L
Packaging:
Amber Glass Bottle
Cubitainer
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R783200020F 20 L Cubitainer
R7832000500 500 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R783200020F Supplier Ricca Chemical Company Supplier No. 78325
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7732-18-5 , 7757-83-7
Molecular Formula Na2O3S
Molecular Weight (g/mol) 126.037
InChI Key GEHJYWRUCIMESM-UHFFFAOYSA-L
Synonym sodium sulfite, disodium sulfite, sodium sulphite, sodium sulfite anhydrous, sulfurous acid, disodium salt, sulftech, natrium sulfurosum, sodiumsulfite, natriumsulfid, natriumsulfit
PubChem CID 24437
ChEBI CHEBI:86477
IUPAC Name disodium;sulfite
SMILES [O-]S(=O)[O-].[Na+].[Na+]

Specifications

Color Colorless
Physical Form Liquid
Quantity 20 L
Assay Percent Range 1.000 ± 0.001 M at 20°C
Solubility Information Solubility in Water: Infinite
Grade Laboratory
Odor Odorless
Packaging Cubitainer
Chemical Name or Material Sodium Sulfite, 1.00 Molar, 2.00 Normal
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.