Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Sulfite, ≥98.0%, ACS Reagent, Solstice

Catalog No. 60045500 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-045-500 500 g Poly Bottle
60-045-499 2.5 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-045-500 Supplier Solstice Supplier No. 239321500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ACS Reagent,≥98.0%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7757-83-7
Molecular Formula Na2O3S
Molecular Weight (g/mol) 126.037
MDL Number MFCD00003503
InChI Key GEHJYWRUCIMESM-UHFFFAOYSA-L
Synonym sodium sulfite, disodium sulfite, sodium sulphite, sodium sulfite anhydrous, sulfurous acid, disodium salt, sulftech, natrium sulfurosum, sodiumsulfite, natriumsulfid, natriumsulfit
PubChem CID 24437
ChEBI CHEBI:86477
IUPAC Name disodium;sulfite
SMILES [O-]S(=O)[O-].[Na+].[Na+]

Specifications

Melting Point 600°C
Color White
Quantity 500 g
Linear Formula Na2SO3
UN Number NONH for all modes of transport
Solubility Information Soluble in water, 126g/L at 20°C
Formula Weight 126.04g/mol
Percent Purity ≥98.0%
Grade ACS Reagent
Packaging Poly Bottle
Appearance Powder or Crystals
Density 2.6g/cm3
Chemical Name or Material Sodium Sulfite
Show More Show Less
EINECSNumber 231-821-4
RTECSNumber WE2150000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.