Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium sulfite EMPROVE™ ESSENTIAL Ph Eur, BP, NF, E 221, 100.5%, MilliporeSigma™

Catalog No. M1066529029 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1066529029 25 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1066529029 Supplier MilliporeSigma Supplier No. 1066529029

Chemical Identifiers

CAS 7757-83-7
Molecular Formula Na2O3S
Molecular Weight (g/mol) 126.04

Specifications

Melting Point > 500°C (decomposition)
pH 8.8 to 10 (50 g/L, H2O, 20°C)
Quantity 25 kg
Assay Percent Range Assay (iodometric): 95.0 to 100.5%; Assay (SO₂): ≥ 48.0%
Solubility Information 220 g/L
Packaging Double PE Sack
Density 2.63 g/cm3 (at 20°C); Bulk Density: 1480 kg/m3
Recommended Storage Store at 5 to 30°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.