Learn More
Hach Company Sodium Sulfite 100 g
Catalog No. 501995901 Shop All Hach Company Products
Encompass
Sodium Sulfite Micro Grade. 100 g.For Zero point DO calibration
Catalog No. 50-199-5901 Supplier Hach Company Supplier No. 2386026
Sodium Sulfite Micro Grade. 100 g.For Zero point DO calibration