Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Sulfite, Purified, Reagents

Catalog No. 01336956
Change view
Click to view available options
Quantity:
500 g
10 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-336-956 10 kg
01-336-954 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-336-956 Supplier Reagents Holdings Llc Supplier No. C232650010KG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sodium Sulfite, Purified, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7757-83-7
Molecular Formula H14Na2O10S
Molecular Weight (g/mol) 252.14
InChI Key WMJRPJZQQSSDBU-UHFFFAOYSA-L
Synonym Sulfurous Acid disodium salt
IUPAC Name disodium heptahydrate sulfite
SMILES O.O.O.O.O.O.O.[Na+].[Na+].[O-]S([O-])=O

Specifications

Melting Point >600°C
Color White
Physical Form Solid
Quantity 10 kg
Grade Pure
Packaging Poly Pail
Specific Gravity 2.633
Chemical Name or Material Sodium Sulfite, Purified
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.