Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Sulfite, Reagent, ACS, 98%, Spectrum™ Chemical
SDP

Catalog No. 18606859 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-859 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-606-859 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S1475500GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

S1475, 7757-83-7, Na2SO3

Spectrum™ Chemical Sodium Sulfite, Reagent, ACS is a used for preserving meat and prevent discoloration of dried fruit. It is a soluble sodium salt of sulfuric acid. It is also used in the paper processing industry and to purify TNT. As an ACS grade Reagent, Spectrum Chemical manufactured compound is used as the quality standard against which other substances are grade and has met the toughest regulatory standards for quality and pureness.

Chemical Identifiers

CAS 7757-83-7
Molecular Formula H14Na2O10S
Molecular Weight (g/mol) 252.14
InChI Key WMJRPJZQQSSDBU-UHFFFAOYSA-L
IUPAC Name disodium heptahydrate sulfite
SMILES O.O.O.O.O.O.O.[Na+].[Na+].[O-]S([O-])=O

Specifications

CAS Max % 1
Physical Form Crystalline Powder or Needles
Quantity 500 g
Percent Purity 98%
Grade ACS/Reagent
Packaging Poly Bottle
Chemical Name or Material Sodium Sulfite
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.