Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Tartrate, 25% w/v, pH 7, Reagents

Sodium Tartrate, 25% w/v, pH 7, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

$60.75

Chemical Identifiers

CAS 7732-18-5 , 6106-24-7
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
IUPAC Name water
SMILES O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
01-333-947
View Documents
Reagents Holdings Llc
CS0132001A
1 L
Each of 1 for $60.75
Add to Cart
 
 Restricted Item
Description

Description

Specifications

Chemical Identifiers

7732-18-5 , 6106-24-7
18.02
water
H2O
XLYOFNOQVPJJNP-UHFFFAOYSA-N
O

Specifications

Colorless
102°C
XLYOFNOQVPJJNP-UHFFFAOYSA-N
water
25%
Poly Bottle
7
Liquid
7732-18-5 , 6106-24-7
H2O
O
18.02
Reagent
1.1352
Non-sterile
Sodium Tartrate, 25% w/v, pH 7
Videos
SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.