Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Tartrate, 25% w/v, pH 7, Reagents

Sodium Tartrate, 25% w/v, pH 7, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Supplier:  Reagents Holdings Llc CS0132001A

Quantity 1 L

 View more versions of this product

Catalog No. 01-333-947


Add to Cart

Description

Description

Specifications

Chemical Identifiers

7732-18-5 , 6106-24-7
18.02
water
H2O
XLYOFNOQVPJJNP-UHFFFAOYSA-N
O

Specifications

Colorless
102°C
XLYOFNOQVPJJNP-UHFFFAOYSA-N
water
25%
Poly Bottle
7
Non-sterile
Sodium Tartrate, 25% w/v, pH 7
7732-18-5 , 6106-24-7
H2O
O
18.02
Reagent
1.1352
1 L
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.