CAS RN 6106-24-7
HYDRANAL™-Sodium tartrate dihydrate, Solstice
Test substance for volumetric Karl Fischer titration (water content ∼15.66%, exact value on report of analysis)
Sodium L-(+)-Tartrate Dihydrate (Crystalline/Certified ACS), Fisher Chemical™
CAS: 6106-24-7 Molecular Formula: C4H8Na2O8 Molecular Weight (g/mol): 230.08 MDL Number: MFCD00150035 InChI Key: FGJLAJMGHXGFDE-UDMGOBQRNA-L Synonym: unii-dia7c37aow,disodium tartrate dihydrate,disodium l-+-tartrate dihydrate,dia7c37aow,disodium +-tartrate dihydrate,sodium l-+-tartrate dihydrate,butanedioic acid, 2,3-dihydroxy-2r,3r-, disodium salt, dihydrate,disodium tartarate dihydrate,sodium tartrate nf,sodium tartrate dihydrate mi PubChem CID: 131855972 IUPAC Name: disodium (2R,3R)-2,3-dihydroxybutanedioate dihydrate SMILES: O.O.[Na+].[Na+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O
Sodium Tartrate Dihydrate (Colorless Crystals), Fisher BioReagents
CAS: 6106-24-7 | C4H8Na2O8 | 230.08 g/mol
Sodium L-(+)-tartrate dihydrate, 99%
CAS: 6106-24-7 Molecular Formula: C4H8Na2O8 Molecular Weight (g/mol): 230.08 MDL Number: MFCD00150035 InChI Key: FGJLAJMGHXGFDE-UDMGOBQRNA-L Synonym: unii-dia7c37aow,disodium tartrate dihydrate,disodium l-+-tartrate dihydrate,dia7c37aow,disodium +-tartrate dihydrate,sodium l-+-tartrate dihydrate,butanedioic acid, 2,3-dihydroxy-2r,3r-, disodium salt, dihydrate,disodium tartarate dihydrate,sodium tartrate nf,sodium tartrate dihydrate mi PubChem CID: 131855972 IUPAC Name: (2R,3R)-2,3-dihydroxybutanedioic acid;sodium;dihydrate SMILES: O.O.[Na+].[Na+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O
Sodium L-(+)-tartrate dihydrate, ACS, 99.0-101.0%
CAS: 6106-24-7 Molecular Formula: C4H8Na2O8 Molecular Weight (g/mol): 230.08 MDL Number: MFCD00150035 InChI Key: FGJLAJMGHXGFDE-UDMGOBQRNA-L Synonym: unii-dia7c37aow,disodium tartrate dihydrate,disodium l-+-tartrate dihydrate,dia7c37aow,disodium +-tartrate dihydrate,sodium l-+-tartrate dihydrate,butanedioic acid, 2,3-dihydroxy-2r,3r-, disodium salt, dihydrate,disodium tartarate dihydrate,sodium tartrate nf,sodium tartrate dihydrate mi PubChem CID: 131855972 SMILES: O.O.[Na+].[Na+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O
L(+)-Tartaric acid, disodium salt, dihydrate, 99+%, ACS reagent
CAS: 6106-24-7 Molecular Formula: C4H8Na2O8 Molecular Weight (g/mol): 230.08 MDL Number: MFCD00150035 InChI Key: FGJLAJMGHXGFDE-UDMGOBQRNA-L Synonym: unii-dia7c37aow,disodium tartrate dihydrate,disodium l-+-tartrate dihydrate,dia7c37aow,disodium +-tartrate dihydrate,sodium l-+-tartrate dihydrate,butanedioic acid, 2,3-dihydroxy-2r,3r-, disodium salt, dihydrate,disodium tartarate dihydrate,sodium tartrate nf,sodium tartrate dihydrate mi PubChem CID: 131855972 SMILES: O.O.[Na+].[Na+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O
Sodium Tartrate, Dihydrate, Purified, Spectrum™ Chemical
CAS: 6106-24-7 Molecular Formula: C4H8Na2O8 Molecular Weight (g/mol): 230.08 MDL Number: MFCD00150035 InChI Key: FGJLAJMGHXGFDE-UDMGOBQRNA-L IUPAC Name: disodium (2R,3R)-2,3-dihydroxybutanedioate dihydrate SMILES: O.O.[Na+].[Na+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O
Sodium Tartrate, Dihydrate, Crystal, Reagent, ACS, 99-101%, Spectrum™ Chemical
CAS: 6106-24-7 Molecular Formula: C4H8Na2O8 Molecular Weight (g/mol): 230.08 MDL Number: MFCD00150035 InChI Key: FGJLAJMGHXGFDE-UDMGOBQRNA-L IUPAC Name: disodium (2R,3R)-2,3-dihydroxybutanedioate dihydrate SMILES: O.O.[Na+].[Na+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O
Sodium Tartrate Dihydrate, Pharmaceutical Secondary Standard, Certified Reference Material, MilliporeSigma™ Supelco™
Certified pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to pharmacopeia primary standards
Sodium Tartrate, Dihydrate, 99.0 to 101.0%, ACS, MilliporeSigma™
CAS: 6106-24-7 Molecular Formula: C4H8Na2O8 Molecular Weight (g/mol): 230.08 MDL Number: MFCD00150035 InChI Key: FGJLAJMGHXGFDE-UDMGOBQRNA-L Synonym: unii-dia7c37aow,disodium tartrate dihydrate,disodium l-+-tartrate dihydrate,dia7c37aow,disodium +-tartrate dihydrate,sodium l-+-tartrate dihydrate,butanedioic acid, 2,3-dihydroxy-2r,3r-, disodium salt, dihydrate,disodium tartarate dihydrate,sodium tartrate nf,sodium tartrate dihydrate mi PubChem CID: 131855972 IUPAC Name: disodium (2R,3R)-2,3-dihydroxybutanedioate dihydrate SMILES: O.O.[Na+].[Na+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O
Sodium Tartrate, Dihydrate, Crystal, Reagent, ACS, 99-101%, Spectrum™ Chemical
CAS: 6106-24-7 Molecular Formula: C4H8Na2O8 Molecular Weight (g/mol): 230.08 MDL Number: MFCD00150035 InChI Key: FGJLAJMGHXGFDE-UDMGOBQRNA-L IUPAC Name: disodium (2R,3R)-2,3-dihydroxybutanedioate dihydrate SMILES: O.O.[Na+].[Na+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O
Sodium Tartrate, Dihydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
Molecular Formula: C4H8Na2O8 Molecular Weight (g/mol): 230.08 MDL Number: MFCD00150035 InChI Key: FGJLAJMGHXGFDE-UDMGOBQRNA-L PubChem CID: 131855972 SMILES: O.O.[Na+].[Na+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O
Sodium Tartrate Dihydrate, ACS Reagent Grade, Ricca Chemical
CAS: 6106-24-7 Molecular Formula: C4H8Na2O8 Molecular Weight (g/mol): 230.08 MDL Number: MFCD00150035 InChI Key: FGJLAJMGHXGFDE-UDMGOBQRNA-L Synonym: unii-dia7c37aow,disodium tartrate dihydrate,disodium l-+-tartrate dihydrate,dia7c37aow,disodium +-tartrate dihydrate,sodium l-+-tartrate dihydrate,butanedioic acid, 2,3-dihydroxy-2r,3r-, disodium salt, dihydrate,disodium tartarate dihydrate,sodium tartrate nf,sodium tartrate dihydrate mi PubChem CID: 131855972 IUPAC Name: disodium (2R,3R)-2,3-dihydroxybutanedioate dihydrate SMILES: O.O.[Na+].[Na+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O
Disodium L-(+)-Tartrate Dihydrate 98.0+%, TCI America™
CAS: 6106-24-7 Molecular Formula: C4H8Na2O8 Molecular Weight (g/mol): 230.08 MDL Number: MFCD00150035 InChI Key: FGJLAJMGHXGFDE-UDMGOBQRNA-L Synonym: unii-dia7c37aow,disodium tartrate dihydrate,disodium l-+-tartrate dihydrate,dia7c37aow,disodium +-tartrate dihydrate,sodium l-+-tartrate dihydrate,butanedioic acid, 2,3-dihydroxy-2r,3r-, disodium salt, dihydrate,disodium tartarate dihydrate,sodium tartrate nf,sodium tartrate dihydrate mi PubChem CID: 131855972 IUPAC Name: disodium (2R,3R)-2,3-dihydroxybutanedioate dihydrate SMILES: O.O.[Na+].[Na+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O
- 1
- 2