Learn More
Apexbio Technology LLC Sodium tartrate 868-18-8 5g
SDP
Catalog No. 502240651
Encompass_Preferred
Sodium tartrate is a small-molecule inhibitor targeting acidic phosphatase It is designed to inhibit acidic phosphatase activity thereby regulating intracellular signaling pathways and enzymes involved in oxidative stress processes Sodium tartrate exerts its biological activity primarily through enzyme inhibition with antioxidant activity In experimental studies sodium tartrate demonstrates modulation of hydrolysis rates in lipid-containing systems at elevated temperatures Based on these pharmacological properties sodium tartrate holds research potential in studies related to oxidative stress enzyme inhibition antioxidant mechanisms and lipid stability in biomedical research
This item is not returnable. View return policy
Catalog No. 50-224-0651 Supplier Apexbio Technology LLC Supplier No. B78445000
Sodium tartrate is a small-molecule inhibitor targeting acidic phosphatase It is designed to inhibit acidic phosphatase activity thereby regulating intracellular signaling pathways and enzymes involved in oxidative stress processes Sodium tartrate exerts its biological activity primarily through enzyme inhibition with antioxidant activity In experimental studies sodium tartrate demonstrates modulation of hydrolysis rates in lipid-containing systems at elevated temperatures Based on these pharmacological properties sodium tartrate holds research potential in studies related to oxidative stress enzyme inhibition antioxidant mechanisms and lipid stability in biomedical research