Learn More
Apexbio Technology LLC Sodium tartrate 868-18-8 10g
SDP
Catalog No. 5000091185
Encompass_Preferred
Sodium tartrate is a small-molecule inhibitor targeting acidic phosphatase It is designed to inhibit acidic phosphatase activity thereby regulating intracellular signaling pathways and enzymes involved in oxidative stress processes Sodium tartrate exerts its biological activity primarily through enzyme inhibition with antioxidant activity In experimental studies sodium tartrate demonstrates modulation of hydrolysis rates in lipid-containing systems at elevated temperatures Based on these pharmacological properties sodium tartrate holds research potential in studies related to oxidative stress enzyme inhibition antioxidant mechanisms and lipid stability in biomedical research
Catalog No. 50-000-91185 Supplier Apexbio Technology LLC Supplier No. B784410000
Sodium tartrate is a small-molecule inhibitor targeting acidic phosphatase It is designed to inhibit acidic phosphatase activity thereby regulating intracellular signaling pathways and enzymes involved in oxidative stress processes Sodium tartrate exerts its biological activity primarily through enzyme inhibition with antioxidant activity In experimental studies sodium tartrate demonstrates modulation of hydrolysis rates in lipid-containing systems at elevated temperatures Based on these pharmacological properties sodium tartrate holds research potential in studies related to oxidative stress enzyme inhibition antioxidant mechanisms and lipid stability in biomedical research