Learn More
Medchemexpress LLC Sodium tartrate | 868-18-8 | MFCD00012468 | 99.0% | 194.05 | C4H4Na2O6 | 5 MG
SDP
Catalog No. 5000264354
Encompass_Preferred
Sodium tartrate is an analytical standard used in research and analytical chemistry. It functions as a pH-regulating agent and antioxidant, and is supplied in small pack sizes suitable for assay and reference use.
  • Serves as an analytical standard for qualitative and quantitative assays.
  • Suitable for HPLC, GC, and MS applications.
  • Acts as a pH-regulating agent and antioxidant in formulations.
  • Provided in small pack sizes for assay and reference use.
  • Molecular weight 194.05 and formula C4H4Na2O6 for precise calculations.
Catalog No. 50-002-64354 Supplier Medchemexpress LLC Supplier No. HY1284765MG
Sodium tartrate is an analytical standard used in research and analytical chemistry. It functions as a pH-regulating agent and antioxidant, and is supplied in small pack sizes suitable for assay and reference use.
  • Serves as an analytical standard for qualitative and quantitative assays.
  • Suitable for HPLC, GC, and MS applications.
  • Acts as a pH-regulating agent and antioxidant in formulations.
  • Provided in small pack sizes for assay and reference use.
  • Molecular weight 194.05 and formula C4H4Na2O6 for precise calculations.