Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sodium taurocholate | 145-42-6 | MFCD00078059 | 97.9% | 537.68 | C26H44NNaO7S | 10 MG
SDP

Catalog No. 5000297271
Encompass_Preferred

Taurocholic acid sodium (sodium taurocholate) analytical standard intended for research and analytical applications. It serves as a reference standard for qualitative, quantitative, and methodological experiments using HPLC, GC, and MS, and has reported bioactivity including immunoregulatory effects and inhibitory potential against hepatic artery ligation-induced biliary damage.

  • Analytical standard for reference and assay purposes.
  • Purity 97.91% (per certificate of analysis).
  • Molecular weight 537.68; CAS number 145-42-6.
  • White to off-white solid, suitable for HPLC, GC, and MS applications.
  • Offered in small reference package sizes, for example 10 mg.

Catalog No. 50-002-97271 Supplier Medchemexpress LLC Supplier No. HYN0545R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Taurocholic acid sodium (sodium taurocholate) analytical standard intended for research and analytical applications. It serves as a reference standard for qualitative, quantitative, and methodological experiments using HPLC, GC, and MS, and has reported bioactivity including immunoregulatory effects and inhibitory potential against hepatic artery ligation-induced biliary damage.

  • Analytical standard for reference and assay purposes.
  • Purity 97.91% (per certificate of analysis).
  • Molecular weight 537.68; CAS number 145-42-6.
  • White to off-white solid, suitable for HPLC, GC, and MS applications.
  • Offered in small reference package sizes, for example 10 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.