Learn More
Selleck Chemical LLC Taurocholic acid sodium salt hydrate S5130-25mg
SDP
Catalog No. 503109758
Encompass_Preferred
Taurocholic acid (Sodium taurocholate hydrate) a bile salt formed in the liver is the product of conjugation of cholic acid with taurine that is involved in the emulsification of lipids Its sodium salt is the chief ingredient of the bile of carnivorous animals Taurocholic acid sodium salt hydrate can be used to induce animal models of Severe Acute Pancreatitis (SAP)
Catalog No. 50-310-9758 Supplier Selleck Chemical LLC Supplier No. S513025MG
Taurocholic acid (Sodium taurocholate hydrate) a bile salt formed in the liver is the product of conjugation of cholic acid with taurine that is involved in the emulsification of lipids Its sodium salt is the chief ingredient of the bile of carnivorous animals Taurocholic acid sodium salt hydrate can be used to induce animal models of Severe Acute Pancreatitis (SAP)