Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sodium tellurite

Catalog No. 40068850G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-068-850G 50 g
40-068-810G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 40-068-850G Supplier Sigma Aldrich Supplier No. 40068850G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Sodium terllurite is an alkali tellurite that is majorly used in the formation of glasses due to its unique properties which include good transmittance, refractive index, and high chemical resistance. It can be used as a tellurite source for the preparation of quantum dots.

Specifications

CAS 10102-20-2
Quantity 50 g
Molecular Formula Na2O3Te
Linear Formula Na2TeO3
MDL Number MFCD00036141
Synonym Disodium trioxotellurate
Molecular Weight (g/mol) 221.58
Percent Purity 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.