Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Tellurite Pentahydrate, TCI America™
SDP

Catalog No. S096525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S096525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S096525G Supplier TCI America Supplier No. S096525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 22451-06-5
Molecular Formula Na2O3Te
MDL Number MFCD00671085

Specifications

Melting Point 300°C
Color White
Quantity 25 g
UN Number 3284
Formula Weight 221.58
Physical Form Crystalline Powder
Chemical Name or Material Sodium Tellurite Pentahydrate
EINECSNumber (1)-0511
RTECSNumber WY2520000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.