Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sodium Tert-Butoxide

Catalog No. 70378825G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-378-825G 25 g
70-378-85G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 70-378-825G Supplier Sigma Aldrich Supplier No. 70378825G
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Sodium tert-butoxide can be used as:A promoter for the synthesis of biaryls by C-H bond arylation of aromatic compounds with haloarenes.A base in palladium catalyzed amination reactions.A base in the synthesis of aryl tert-butyl ethers from unactivated aryl halides in presence of palladium catalyst.In the desulfonylation of N-indoles and N-azaindoles.As a base in the synthesis of sodium acrylates from olefins and CO2.

Specifications

CAS 865-48-5
Molecular Formula C4H9NaO
Linear Formula NaOC(CH3)3
MDL Number MFCD00040575
Quantity 25 g
Molecular Weight (g/mol) 96.1 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.