Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sodium Tert-Butoxide

Catalog No. 3592705G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
1.5 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
35-927-05G 5 g
35-927-015KG 1.5 kg
35-927-0500G 500 g
35-927-025G 25 g
35-927-0100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Danger
Catalog No. 35-927-05G Supplier Sigma Aldrich Supplier No. 3592705G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Sodium tert-butoxide may be used as a safe and effective alternative to sodium hydride for the coupling of hydroxyethyl adenine with diethyl p-toluenesulfonyloxymethanephosphonate to form 9-[2-(diethylphosphonomethoxy)ethyl]adenine (diethyl-PMEA), which is a key intermediate to prepare adefovir dipivoxil.It can also be used:To facilitate the coupling of aryl halides with benzene derivativesin the presence of 10-phenanthroline derivative.To activate first-row transition-metal pre-catalysts.As a base for the amination of aryl chlorides in the presence of a novel palladacyclic precatalyst.

Specifications

CAS 865-48-5
Molecular Formula C4H9NaO
Linear Formula NaOC(CH3)3
MDL Number MFCD00040575
Quantity 5 g
Molecular Weight (g/mol) 96.1 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.