Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sodium Tert-Butoxide Solution

Catalog No. 7027064X2ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
4 x 2 mL
100 mL
4 x 25 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-270-64X2ML 4 x 2 mL
70-270-6100ML 100 mL
70-270-6 4 x 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 70-270-64X2ML Supplier Sigma Aldrich Supplier No. 7027064X2ML
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

ZerO2 products-ßare rigorously degassed with highly pure inert gas providing solvents and solutions-ß(anhydrous if specified)-ßwith very low residual oxygen content.ZerO2 is a registered trademark of Sigma-Aldrich Co. LLC

Specifications

CAS 865-48-5
Density 0.911 g/mL at 25°C
Molecular Formula C4H9NaO
Linear Formula C4H9NaO
Refractive Index n20/D 1.413
MDL Number MFCD00040575
Quantity 4 x 2 mL
Synonym Degassed and low oxygen sodium tert-butoxide solution
Molecular Weight (g/mol) 96.1 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.