Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Tetraborate Decahydrate (Crystalline/Certified ACS), Fisher Chemical™

Catalog No. S24810 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 g
3 kg
10 kg
Packaging:
Poly Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S248-10 10 kg Poly Pail
S248-500 500 g Poly Bottle
S248-3 3 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S248-10
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

Chemical Identifiers

CAS 1303-96-4
Molecular Formula B4Na2O7 · 10 H2O
MDL Number MFCD00149193
Synonym Sodium Biborate, Sodium Pyroborate, Borax

Specifications

Melting Point 75°C
Color White
pH 9
Physical Form Powder/Solid
Quantity 10 kg
Assay Percent Range 99.5 to 105.0 %
Formula Weight 381.37
Percent Purity 99.5 to 105.0%
Grade Certified ACS
Identification Pass Test
Packaging Poly Pail
Density 1.7300g/cm³
Chemical Name or Material Sodium Tetraborate Decahydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.