Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium tetraborate decahydrate, Puriss., meets analytical spec. of BP, NF, Ph. Eur., 99.0-103.0%, Solstice

Catalog No. 6001910 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
2.5 kg
6 x 2.5 kg
Packaging:
Poly bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-019-10 2.5 kg Poly bottle
60-044-704 6 x 2.5 kg Poly bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-019-10 Supplier Solstice Supplier No. 116252.5KG
Add to Cart
Add to Cart

Puriss., meets analytical specification of BP, NF, Ph. Eur., 99.0-103.0%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1303-96-4
MDL Number MFCD00149193

Specifications

Quantity 2.5 kg
Linear Formula Na2B4O7 · 10H2O
UN Number NONH for all modes of transport
Formula Weight 381.37g/mol
Percent Purity 99 to 103.0%
Grade puriss.
Packaging Poly bottle
Density 1.73g/mL at 25°C
Chemical Name or Material Sodium Tetraborate Decahydrate, Meets analytical specification of Ph. Eur.BP, NF
Hazard Statement P201-P308 + P313
Precautionary Statement H360FD
EINECSNumber 215-540-4
RTECSNumber VZ2275000
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.