Learn More
Sodium tetraborate decahydrate, Puriss., meets analytical spec. of BP, NF, Ph. Eur., 99.0-103.0%, Solstice
Shop All Solstice Research Chemicals ProductsDescription
- Leading the Industry For Over 65 Years
- Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Boiling Point | 1,575°C |
| CAS Max % | 100.0000% |
| Physical Form | Crystalline Powder |
| Quantity | 6 x 2.5 kg |
| Assay Percent Range | 99 to 103.0% |
| Linear Formula | Na2B4O7 · 10H2O |
| UN Number | NONH for all modes of transport |
| Molecular Weight (g/mol) | 381.37 |
| Formula Weight | 381.37 |
| Percent Purity | 99.0 to 103.0% |
| Show More |
Safety and Handling
| Hazard Statement | P201-P308 + P313 |
| Precautionary Statement | H360FD |
| ShelfLife | 1260 days from date of manufacture |
| EINECSNumber | 215-540-4 |
| RTECSNumber | VZ2275000 |
| Recommended Storage | Room Temp |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.