Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Tetraborate Decahydrate (White Crystals/Molecular Biology), Fisher BioReagents

Catalog No. BP175500 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP175-500 500 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP175-500

CAS: 1303-96-4 | B4Na2O7 · 10 H2O

Glass Bottle; 500g

Sodium Borate can be used in the preparation of TBE electrophoresis buffer.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium Tetraborate Decahydrate
Melting Point 75°C
Color White
CAS 1303-96-4, 1330-43-4
pH 9
Physical Form Powder/Solid
Quantity 500 g
Assay Percent Range 99.5 to 105.0 %
Molecular Formula B4Na2O7 · 10 H2O
Merck Index 15, 8726
Synonym Borax, Boric Acid, Disodium Salt
Formula Weight 381.36
Percent Purity 99.5 to 105.0%
Grade Molecular Biology
Identification Pass Test
Packaging Glass Bottle
Density 1.7300g/cm³
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.