Learn More
Strem, An Ascensus Company CAS# 13820-53-6. 10g. Sodium tetrachloropalladate(II) trihydrate, 99%. MFCD00149171
SDP
Catalog No. 501385560
Encompass
CAS# 13820-53-6. 10g. Sodium tetrachloropalladate(II) trihydrate, 99%. MFCD00149171. Molecular Weight: 294.19 (348.22). Molecular Formula: Na2PdCl4∙3H2O. Color/form: reddish-brown pwdr. Strem# 93-4613. http://www.strem.com/catalog/v/93-4613/
Catalog No. 50-138-5560 Supplier Strem, An Ascensus Company Supplier No. 93461310G
CAS# 13820-53-6. 10g. Sodium tetrachloropalladate(II) trihydrate, 99%. MFCD00149171. Molecular Weight: 294.19 (348.22). Molecular Formula: Na2PdCl4∙3H2O. Color/form: reddish-brown pwdr. Strem# 93-4613. http://www.strem.com/catalog/v/93-4613/