Learn More
Sodium tetrachloroaurate hydrate, 98%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Brown to Yellow |
| Physical Form | Lump/Powder |
| Quantity | 1 g |
| Assay Percent Range | 48.3 to 50.3% (Au) |
| Linear Formula | NaAuCl4·xH2O |
| Solubility Information | Solubility in water: soluble. Other solubilities: soluble in alcohol and ether |
| Formula Weight | 361.77 |
| Percent Purity | 98% |
| Packaging | Glass bottle |
| Chemical Name or Material | Sodium tetrachloroaurate hydrate |
Safety and Handling
| missing translation for 'healthHazard1' | GHS Signal Word: Warning |
| missing translation for 'healthHazard2' | GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. Harmful if swallowed. May be corrosive to metals. |
| missing translation for 'healthHazard3' | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Call a POISON CENTER or doctor |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.