Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium tetrachloroaurate hydrate, 98%

Catalog No. AC396270050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC396270050 5 g Glass bottle
AC396270010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC396270050 Supplier Thermo Scientific Chemicals Supplier No. 396270050
Add to Cart

CAS: 29156-65-8 | AuCl4Na | 361.76 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29156-65-8
Molecular Formula AuCl4Na
Molecular Weight (g/mol) 361.76
MDL Number MFCD00149161
InChI Key IXPWAPCEBHEFOV-UHFFFAOYSA-J
Synonym sodium tetrachloroaurate iii hydrate, aucl4.na.h2o, sodium tetrachloroaurate 1--water 1/1/1
PubChem CID 18410391
IUPAC Name sodium;tetrachlorogold(1-);hydrate
SMILES [Na+].Cl[Au-](Cl)(Cl)Cl

Specifications

Color Brown to Yellow
Physical Form Lump/Powder
Quantity 5 g
Assay Percent Range 48.3 to 50.3% (Au)
Linear Formula NaAuCl4·xH2O
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol and ether
Formula Weight 361.77
Percent Purity 98%
Packaging Glass bottle
Chemical Name or Material Sodium tetrachloroaurate hydrate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
May be corrosive to metals.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.