Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sodium tetrachloropalladate(II) | 13820-53-6 | 294.21 | 250 MG
SDP

Catalog No. 5000450085
Encompass_Preferred

Sodium tetrachloropalladate(II) is an organic compound primarily utilized as a catalyst and a raw material in dye production. It facilitates or promotes various chemical reactions, finding extensive applications in organic synthesis and dye preparation. Additionally, it is employed for metallization, silver plating, and electroplating of circuit boards. For in vitro applications, it serves as a biochemical reagent for life science research.

  • Used as a catalyst and raw material in dye production
  • Facilitates and promotes various chemical reactions
  • Extensive applications in organic synthesis and dye preparation
  • Employed for metallization, silver plating, and electroplating of circuit boards
  • Biochemical reagent for life science research
  • Brown to reddish brown solid appearance
  • Purity of ≥99.0%
  • Molecular weight of 294.21
  • Recommended storage at 4°C, sealed and away from moisture

Catalog No. 50-004-50085 Supplier Medchemexpress LLC Supplier No. 250MGHYW008145
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sodium tetrachloropalladate(II) is an organic compound primarily utilized as a catalyst and a raw material in dye production. It facilitates or promotes various chemical reactions, finding extensive applications in organic synthesis and dye preparation. Additionally, it is employed for metallization, silver plating, and electroplating of circuit boards. For in vitro applications, it serves as a biochemical reagent for life science research.

  • Used as a catalyst and raw material in dye production
  • Facilitates and promotes various chemical reactions
  • Extensive applications in organic synthesis and dye preparation
  • Employed for metallization, silver plating, and electroplating of circuit boards
  • Biochemical reagent for life science research
  • Brown to reddish brown solid appearance
  • Purity of ≥99.0%
  • Molecular weight of 294.21
  • Recommended storage at 4°C, sealed and away from moisture

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.