Learn More
Sodium tetrafluoroborate, 97%
Description
Sodium tetrafluoroborate is used in fluxes, metal finishing and in plating circuits. It is used as a starting material in the preparation of ionic liquids as well as boron trifluoride. It acts as an efficient catalyst for the synthesis of 3,4-dihydropyrimidin-2(1H)-ones and thiones.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Sodium tetrafluoroborate is used in fluxes, metal finishing and in plating circuits. It is used as a starting material in the preparation of ionic liquids as well as boron trifluoride. It acts as an efficient catalyst for the synthesis of 3,4-dihydropyrimidin-2(1H)-ones and thiones.
Solubility
Soluble in water.
Notes
Hygroscopic and moisture sensitive. Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | 384°C (dec.) |
| Quantity | 500 g |
| Linear Formula | NaBF4 |
| Merck Index | 14,8617 |
| Solubility Information | Soluble in water. |
| Formula Weight | 109.79 |
| Percent Purity | 97% |
| Sensitivity | Hygroscopic |
| Density | 2.47 |
| Chemical Name or Material | Sodium tetrafluoroborate |
Safety and Handling
| Hazard Category | H314 |
| Hazard Statement | GHS H Statement H318-H335-H315 Causes serious eye damage. May cause respiratory irritation. Causes skin irritation. |
| Precautionary Statement | P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
| DOTInformation | Hazard Class: 8; Packaging Group: II |
| EINECSNumber | 237-340-6 |
| RTECSNumber | ED2975000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.