Learn More
Strem, An Ascensus Company CAS# 79060-88-1. 250mg. Sodium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate, min. 98% NaBARF. MFCD00043323
SDP
Catalog No. 5090109458
Encompass
CAS# 79060-88-1. 250mg. Sodium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate, min. 98% NaBARF. MFCD00043323. Molecular Weight: 886.20. Molecular Formula: Na+[B(C6H3(CF3)2)4]ae. Color/form: tan pwdr. Strem# 11-0590. http://www.strem.com/catalog/v/11-0590/
Catalog No. 50-901-09458 Supplier Strem, An Ascensus Company Supplier No. 110590250MG
Add to Cart
Add to Cart
CAS# 79060-88-1. 250mg. Sodium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate, min. 98% NaBARF. MFCD00043323. Molecular Weight: 886.20. Molecular Formula: Na+[B(C6H3(CF3)2)4]ae. Color/form: tan pwdr. Strem# 11-0590. http://www.strem.com/catalog/v/11-0590/