Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sodium Tetrakis(4-Fluorophenyl)Borate Dihydrate

Catalog No. 72016500MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
72-016-500MG 500 mg
72-016-100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 72-016-500MG Supplier Sigma Aldrich Supplier No. 72016500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reagent for the gravimetric determination of cesium

Specifications

CAS 207683-22-5
Molecular Formula C24H16BF4Na -+ 2H2O
Linear Formula (FC6H4)4BNa-+2H2O
MDL Number MFCD00149598
Quantity 500 mg
Synonym Cesibor; Tetrakis(4-fluorophenyl)boron sodium
Molecular Weight (g/mol) 450.21 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.