Learn More
AdipoGen Sodium tetrap-tolylborate
SDP
Catalog No. 502560090
Encompass_Preferred
Chemical. CAS 15738-23-5. Formula C28H28BNa. MW 398.32. Synthetic. Analytical/Chromatography reagent. Ion sensor compound. Additive for ISE.
Catalog No. 50-256-0090 Supplier AdipoGen Supplier No. CDXS0032G010
Chemical. CAS 15738-23-5. Formula C28H28BNa. MW 398.32. Synthetic. Analytical/Chromatography reagent. Ion sensor compound. Additive for ISE.