Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium tetraphenyl borate, ACS, MilliporeSigma™

Catalog No. M1066690025 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Plastic Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1066690025 25 g Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1066690025 Supplier MilliporeSigma Supplier No. 1.06669.0025
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity ACS reagent grade sodium tetraphenylborate suitable for precise analytical applications including cannabis testing.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143-66-8
Molecular Formula C24H20BNa
Molecular Weight (g/mol) 342.22
MDL Number MFCD00011494
InChI Key HFSRCEJMTLMDLI-UHFFFAOYSA-N
Synonym sodium tetraphenylborate, tetraphenylboron sodium, sodium tetraphenylboron, kalignost, sodium tetraphenylborate 1-, kalibor, dotite kalibor, kariporu k, borate 1-, tetraphenyl-, sodium, sodium tetraphenylboride
PubChem CID 2723787
IUPAC Name sodium tetraphenylboranuide
SMILES [Na+].C1=CC=C(C=C1)[B-](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color White
Physical Form Solid
Quantity 25 g
Linear Formula Na[B(C6H5)4]
Solubility Information Soluble (2°C)
Formula Weight 342.22
Percent Purity ≥99.5%
Grade ACS/EP/Reagent
Packaging Plastic Bottle
Density Bulk Density: 500 kg/m3
Chemical Name or Material Sodium Tetraphenylborate, For Analysis
Show More Show Less
Hazard Category SignalWord: Danger
Recommended Storage Store at +15°C to +25°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.