Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sodium Tetraphenylborate

Catalog No. T254021KG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T254021KG 1 kg
T2540225G 25 g
T254025G 5 g
T25402100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Danger
Catalog No. T254021KG Supplier Sigma Aldrich Supplier No. T254021KG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Sodium tetraphenylborate can form hydrophobic salts with amines, ammonium ions, and large alkali metal ions such as cesium or rubidium ions. Hence it is commonly used in gravimetric analysis as a precipitating agent.

Specifications

CAS 34592-47-7
Melting Point 190°C to 200°C
Molecular Formula C4H7NO2S
Linear Formula C4H7NO2S
MDL Number MFCD00005212
Quantity 1 kg
Synonym Tetraphenylboron sodium
Molecular Weight (g/mol) 133.17 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.