Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sodium Tetraphenylborate

Catalog No. 7202050G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
72-020-50G 50 g
72-020-250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 72-020-50G Supplier Sigma Aldrich Supplier No. 7202050G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Sodium tetraphenylborate (Ph4BNa) undergoes rhodium-catalyzed addition reactions with N-phenylsulfonyl aldimines to afford R(Ph)CHNHSO2Ph.Sodium tetraphenylborate (Ph4BNa) may be used in the synthesis of polyfunctional biaryls via modified Suzuki cross-coupling reaction. It may be employed as a derivatization reagent for the analysis of methylmercury and ethylmercury by gas chromatography (GC) coupled with atomic fluorescence spectrometry (AFS), microwave-induced plasma atomic emission spectrometry (AES) and mass spectrometry (MS).Reagent for the precipitation and titration of potassium; For the amperometric titration of amines, quaternary ammonium compounds and potassium; For the titration of quaternary ammonium surfactants; Titrant for amines and NH4Cl in the presence of a K selective electrode

Specifications

CAS 143-66-8
Molecular Formula C24H20BNa
Linear Formula (C6H5)4BNa
MDL Number MFCD00011494
Quantity 50 g
Synonym Tetraphenylboron sodium
Molecular Weight (g/mol) 342.22 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.