Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sodium tetraphenylborate | 143-66-8 | MFCD00011494 | 99.6% | 342.22 g/mol | C24H20BNa | 25 G
SDP

Catalog No. 5000303551
Encompass_Preferred

Sodium tetraphenylborate is a tetraphenylboron salt used as a precipitating agent and laboratory reagent for ion-exchange, separations, and analytical applications. Its high purity and defined solubility profile support reproducible analytical workflows.

  • High purity: 99.6%.
  • Chemical formula C24H20BNa and molecular weight 342.22 g/mol.
  • Soluble in DMSO (200 mg/mL); sparingly soluble in water (3.33 mg/mL with warming).
  • Suitable for precipitating cations and routine analytical precipitations.
  • Available in multiple package sizes for laboratory use.

Catalog No. 50-003-03551 Supplier Medchemexpress LLC Supplier No. 25GHYW025784
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sodium tetraphenylborate is a tetraphenylboron salt used as a precipitating agent and laboratory reagent for ion-exchange, separations, and analytical applications. Its high purity and defined solubility profile support reproducible analytical workflows.

  • High purity: 99.6%.
  • Chemical formula C24H20BNa and molecular weight 342.22 g/mol.
  • Soluble in DMSO (200 mg/mL); sparingly soluble in water (3.33 mg/mL with warming).
  • Suitable for precipitating cations and routine analytical precipitations.
  • Available in multiple package sizes for laboratory use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.