Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Tetraphenylboron (Sodium Tetraphenylborate), Reagents

Catalog No. 01336952
Change view
Click to view available options
Quantity: 25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01-336-952 Quantity: 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-336-952 Supplier Reagents Holdings Llc Supplier No. C232720025C1
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sodium Tetraphenylboron (Sodium Tetraphenylborate), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 143-66-8
Molecular Formula C24H20BNa
Molecular Weight (g/mol) 342.22
MDL Number MFCD00011494
InChI Key HFSRCEJMTLMDLI-UHFFFAOYSA-N
Synonym Sodium tetraphenylborate
IUPAC Name sodium tetraphenylboranuide
SMILES [Na+].C1=CC=C(C=C1)[B-](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color White
Physical Form Solid
Quantity 25 g
Grade Reagent
Packaging Glass Bottle
Specific Gravity 1
Chemical Name or Material Sodium Tetraphenylboron (Sodium Tetraphenylborate)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.