Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Tetraphenylboron TS, Ricca Chemical
SDP

Catalog No. 785416 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
7854-16 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 7854-16 Supplier Ricca Chemical Company Supplier No. 785416
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 143-66-8 , 7732-18-5
Molecular Formula C24H20BNa
Molecular Weight (g/mol) 342.22
MDL Number MFCD00011494
InChI Key HFSRCEJMTLMDLI-UHFFFAOYSA-N
Synonym sodium tetraphenylborate, tetraphenylboron sodium, sodium tetraphenylboron, kalignost, sodium tetraphenylborate 1-, kalibor, dotite kalibor, kariporu k, borate 1-, tetraphenyl-, sodium, sodium tetraphenylboride
PubChem CID 2723787
IUPAC Name sodium tetraphenylboranuide
SMILES [Na+].C1=CC=C(C=C1)[B-](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color Colorless
CAS Min % 97.0, 0.58
CAS Max % 100.0, 0.61
Grade Laboratory
Packaging Amber Glass Bottle
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Sodium Tetraphenylboron, Test Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.